Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepsin Monoclonal Antibody (3H10.1), Functional Grade, In Vivo GOLD™, Leinco Technologies™
SDP

Mouse Monoclonal Antibody

Supplier:  Leinco Technologies Inc H5500100MG

Encompass
Encompass_Preferred

Catalog No. 50-254-4163


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Specificity: Clone 3H10.1.2 is able to recognize full-length native Hepsin expressed on the cell surface in addition to the recombinant soluble form. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

Hepsin Monoclonal antibody specifically detects Hepsin in Human samples. It is validated for ELISA, Flow Cytometry, Functional Assay
Specifications

Specifications

Hepsin
Monoclonal
≥5.0 mg/mL
PBS with no preservative; pH 7.2-7.4
P05981
Mouse
Protein A/G
RUO
3249
4°C short term, -80°C long term
Liquid
ELISA, Flow Cytometry, Functional Assay
3H10.1
Functional Grade
HPN
hepsin; hepsin (transmembrane protease, serine 1); Hlb320; HPN; Serine protease hepsin; Serine protease hepsin catalytic chain; Serine protease hepsin non-catalytic chain; testicular tissue protein Li 85; TMPRSS1; Transmembrane protease serine 1
Human hepsin protein
100 mg
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.