Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepsin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530062 Supplier Invitrogen™ Supplier No. PA530062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, HeLa. Predicted reactivity: Mouse (91%), Rat (91%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Hepsin, encoded by the HPN gene, is a type II transmembrane serine protease prominently expressed in tissues such as the liver, kidney, and prostate. Hepsin plays a crucial role in proteolytic processes involved in extracellular matrix remodeling, cell growth, and development. It is known for activating precursor molecules, including hepatocyte growth factor (HGF), which is essential for liver development and regeneration. Furthermore, hepsin's involvement extends to influencing cell signaling pathways related to tissue homeostasis and repair. In the context of cancer, particularly prostate cancer, hepsin is implicated in tumor progression and metastasis by promoting extracellular matrix degradation, thereby facilitating tumor invasion and spread. Aberrant expression or activity of hepsin is associated with pathophysiological changes in the tumor microenvironment, making it a target for therapeutic interventions aimed at controlling cancer progression. Research continues to explore hepsin's biochemical functions and regulatory mechanisms, providing insights into its potential roles in both normal physiology and disease states.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepsin
Applications Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene HPN
Gene Accession No. P05981
Gene Alias hepsin; hepsin (transmembrane protease, serine 1); Hlb320; HPN; Serine protease hepsin; Serine protease hepsin catalytic chain; Serine protease hepsin non-catalytic chain; testicular tissue protein Li 85; TMPRSS1; Transmembrane protease serine 1
Gene Symbols HPN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human Hepsin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3249
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.