Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heptacosane 97.0+%, TCI America™

Click to view available options
Quantity:
1 g
10 g
Specifications
Specifications
Melting Point | 59°C |
Color | White |
Boiling Point | 270°C |
Quantity | 10 g |
Formula Weight | 380.75 |
Percent Purity | ≥97.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Heptacosane |
Safety and Handling
EINECSNumber | (2)-0010 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction