Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptafluorobutyric Acid, 99.6%, MP Biomedicals™ PROMO

Catalog No. ICN5123725P Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 mL
25 mL

Heptafluorobutyric Acid is used in determination of amino acid sequences in proteins.

  • Heptafluorobutyric Acid is used as a mobile phase modifier which significantly improved selectivity in the HPLC analysis of histone proteins.
  • It can be used at a concentration of 0.1% in the mobile phase of an HPLC/LC-MS protocol for the detection of marine bacterioplankton siderophores.
  • It is used in the chiral LC analysis of (+)- and (-)-epibatidine.
  • The analysis of selenium containing compounds using HFBA in the MS matrix has been described.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4HF7O2
Molecular Weight (g/mol) 214.04
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
PubChem CID 9777
ChEBI CHEBI:39426
IUPAC Name heptafluorobutanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Colorless
Density 1.645 g/mL
Assay Percent Range 99.6%
Quantity 25 mL
Synonym heptafluorobutanoic acid
Formula Weight 214.04
Physical Form Liquid
Chemical Name or Material Heptafluorobutyric Acid

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.