Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptakis-6-iodo-6-deoxy-β-cyclodextrin, Thermo Scientific™

Catalog No. AC299015000
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mg
Packaging:
Glass bottle
Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30754-23-5
Molecular Formula C42H63I7O28
Molecular Weight (g/mol) 1904.28
MDL Number MFCD00803290
InChI Key YODKQJPYQJRHEQ-FOUAGVGXSA-N
Synonym heptakis-6-iodo-6-deoxy-beta-cyclodextrin
PubChem CID 11105283
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CI)CI)CI)CI)CI)CI)O)O)I

Specifications

Melting Point 218°C to 222°C
Color White
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in dmf,dmso,hot methylethyleneglycol
Formula Weight 1904.28
Physical Form Solid
Chemical Name or Material Heptakis-6-iodo-6-deoxy-β-cyclodextrin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.