Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane CHROMASOLV™, for HPLC, ≥99%, Solstice

Catalog No. 6001017 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-010-17 2.5 L
60-010-16 1 L
60-010-22 6 x 1 L
60-010-20 4 x 2.5 L
60-010-21 4 x 4 L
60-010-14 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-010-17 Supplier Solstice Supplier No. 348732.5L

Greener alternatives to Hexane in HPLC.

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -91°C
Density 0.684g/mL at 25°C
Quantity 2.5 L
Refractive Index n20/D 1.387
Linear Formula CH3(CH2)5CH3
Vapor Pressure 83mmHg (37.7°C)
Beilstein 1730763
Boiling Point 98°C
Formula Weight 100.2g/mol
Percent Purity ≥99%
Grade filtration through 0.2μm
Chemical Name or Material Heptane, for HPLC
Show More Show Less
Hazard Statement P210-P273-P301 + P310-P304 + P340 + P312-P331-P391
Precautionary Statement H225-H304-H315-H336-H410
EINECSNumber 205-563-8
RTECSNumber MI7700000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.