Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane, HPLC Grade, Spectrum™ Chemical
SDP

Catalog No. 18603110 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-110 4 x 4 L Amber Glass Bottle
18-603-108 4 L Amber Glass Bottle
18-603-111 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-110 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HP6624LTCS4
Add to Cart
Add to Cart

HP662, 142-82-5, C7H16

Spectrum™ Chemical Heptane, HPLC Grade is a straight-chain alkane applied in laboratories as a non-polar mobile phase carrier solvent in high pressure liquid chromatography. All HPLC solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
IUPAC Name heptane
SMILES CCCCCCC

Specifications

CAS Max % 100%
Quantity 4 x 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 2 ppm
Water 0.02%
UN Number UN1206
Boiling Point 98.4°C
Percent Purity >99%
Grade High Purity
Chemical Name or Material Heptane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.