Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0%, Solstice

Catalog No. 6001034 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-010-34 19 L Metal Can
60-010-35 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-010-34 Supplier Solstice Supplier No. PP20619
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

CAS Max % 100.0000%
Density 0.684 g/cm3
Quantity 19 L
Flash Point −4°C
Packaging Metal Can
Water 0.01% max.
UN Number UN1206
Beilstein 1730763
Boiling Point 98.4°C
Solubility Information 0.03% in Water at 20°C
Formula Weight 100.21
Percent Purity >99%
Grade ACS
Chemical Name or Material Heptane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.