Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane, Puriss. p.a., Reag. Ph. Eur., ≥99% n-heptane basis (GC), Solstice

Catalog No. 60046106 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
6 x 1 L
Packaging:
Aluminum Bottle
Glass Bottle
Not Available
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-106 1 L Glass Bottle
60-046-112 6 x 1 L Not Available
60-046-109 4 x 2.5 L Glass Bottle
60-046-107 2.5 L Glass Bottle
60-046-111 5 L Aluminum Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-046-106 Supplier Solstice Supplier No. 322871L
Add to Cart
Add to Cart

Puriss. p.a., Reag. Ph. Eur.,≥99% n-heptane basis (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -91°C
Appearance Colorless liquid
Density 0.684g/mL at 25°C
Quantity 1 L
Explosion Limit 0.07
Flash Point −4°C (24.8°F)
Ignition Point 433°F
Packaging Glass Bottle
Refractive Index n20/D 1.3870 to 1.3880
Residue after Evaporation 0 to 0.001%
Titratable Acid Free acid (as CH3COOH): ≤0.0005%
Linear Formula CH3(CH2)5CH3
Vapor Pressure 40mmHg (20°C), 83mmHg (37.7°C)
UN Number UN1206
Beilstein 1730763
Boiling Point 98.2°C to 98.4°C
Formula Weight 100.2g/mol
Percent Purity ≥99% n to heptane basis (GC)
Grade puriss. p.a.
Chemical Name or Material Heptane, Reag. Ph. Eur.
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P261-P273-P301 + P310-P331-P501
Precautionary Statement H225-H304-H315-H336-H410
DOTInformation Class 3 - PG 2 - Heptanes
EINECSNumber 205-563-8
RTECSNumber MI7700000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.