Learn More
Heptane, Puriss. p.a., Reag. Ph. Eur., ≥99% n-heptane basis (GC), Solstice
Shop All Honeywell Research Chemicals ProductsDescription
- Solstice has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | -91°C |
| Appearance | Colorless liquid |
| Density | 0.684g/mL at 25°C |
| Quantity | 4 x 2.5 L |
| Explosion Limit | 0.07 |
| Flash Point | −4°C (24.8°F) |
| Ignition Point | 433°F |
| Packaging | Glass Bottle |
| Refractive Index | n20/D 1.3870 to 1.3880 |
| Residue after Evaporation | 0 to 0.001% |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | P210-P261-P273-P301 + P310-P331-P501 |
| Precautionary Statement | H225-H304-H315-H336-H410 |
| DOTInformation | Class 3 - PG 2 - Heptanes |
| EINECSNumber | 205-563-8 |
| RTECSNumber | MI7700000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.