Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptanes, Technical, Reagents

Catalog No. 01337225
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-225 4 L
01-337-227 1 L
01-337-226 20 L
01-337-224 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-225 Supplier Reagents Holdings Llc Supplier No. C22130004C
Add to Cart
Add to Cart

Heptanes, Technical, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym Heptanes (mixed isomers)
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -90.6°C
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.6795
Boiling Point 98.4°C
Grade Technical
Chemical Name or Material Heptanes, Technical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.