Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl nitrite, 97%, stabilized

Catalog No. AC165700025
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass Bottle
Plastic bottle

CAS: 110-46-3 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-46-3, 497-19-8
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00002057
InChI Key OWFXIOWLTKNBAP-UHFFFAOYSA-N
Synonym isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i
PubChem CID 8053
ChEBI CHEBI:2691
IUPAC Name 3-methylbutyl nitrite
SMILES CC(C)CCON=O

Specifications

Color Yellow
Density 0.8700g/mL
Boiling Point 96.0°C to 99.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Refractive Index 1.3840 to 1.3880
Linear Formula (CH3)2CHCH2CH2ONO
Quantity 2.5 L
Beilstein 01, 402
Fieser 01,40; 04,270; 05,358; 06,307; 09,258
Specific Gravity 0.87
Merck Index 15, 5168
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: miscible with alcohol,chloroform and ether
Stabilizer 0.2% sodium carbonate
Formula Weight 117.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Isoamyl nitrite, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-770-8
RTECSNumber NT0187500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.