Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptyl Sulfide 97.0+%, TCI America™
SDP

Catalog No. H037625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H037625G 25 g
H03765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H037625G Supplier TCI America Supplier No. H037625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 629-65-2
Molecular Formula C14H30S
Molecular Weight (g/mol) 230.45
MDL Number MFCD00027315
InChI Key LEMIDOZYVQXGLI-UHFFFAOYSA-N
Synonym Diheptyl Sulfide
PubChem CID 69422
IUPAC Name 1-(heptylsulfanyl)heptane
SMILES CCCCCCCSCCCCCCC

Specifications

Melting Point -11°C
Color Yellow
Boiling Point 298°C
Quantity 25 g
Formula Weight 230.45
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptyl Sulfide
EINECSNumber (2)-0472
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.