Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptylbenzene, Thermo Scientific™

Catalog No. AC150980050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1078-71-3
Molecular Formula C13H20
Molecular Weight (g/mol) 176.303
MDL Number MFCD00009545
InChI Key LBNXAWYDQUGHGX-UHFFFAOYSA-N
Synonym 1-phenylheptane, benzene, heptyl, n-heptylbenzene, heptane, 1-phenyl, heptane, 1 or 7-phenyl, unii-12uc21c7sn, phenylheptane, heptyl-benzene, heptane, phenyl
PubChem CID 14115
IUPAC Name heptylbenzene
SMILES CCCCCCCC1=CC=CC=C1

Specifications

Color Colorless
Density 0.8600g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4840 to 1.4860 (20°C, 589nm)
Linear Formula C6H5(CH2)6CH3
Quantity 5 g
Beilstein 05, 451
Formula Weight 176.3
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Heptylbenzene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.