Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER-2/c-erbB-2/neu Ab-17 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS730P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS730P1 500 μL Concentrated 200μg/mL
MS730P 1 mL Concentrated 200μg/mL
MS730P1ABX 100 μL Concentrated 1mg/mL
MS730PABX 200 μL Concentrated 1mg/mL
MS730R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS730P1 Supplier Epredia Supplier No. MS730P1A
Add to Cart
Add to Cart

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation (For cytoplasmic), Epredia™ HER-2/c-erbB-2/neu Ab-17, Mouse Monoclonal Antibody provides accurate, reproducible results..

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: e2-4001 + 3B5

Isotype: IgG1 + IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Cytoplasmic

Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human HER-2 protein (3B5)

Molecular Weight: 185kDa

Positive Control: LS174T cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (For cytoplasmic)

Specifications

Antigen HER-2/c-erbB-2/neu Ab-17
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone e2-4001 + 3B5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human c-erbB-2 protein (3B5)
Quantity 500 μL
Regulatory Status ASR, IVD
Research Discipline EGFR Family
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.