Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER2 Trastuzumab Biosimilar anti-Human, Unconjugated, Clone: Trastuzumab.rMAb (also known as Trastuzumab Biosimilar), BD Phrmingen™

Catalog No. BDB569599 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569599 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569599 Supplier BD Biosciences Supplier No. 569599
Add to Cart
Add to Cart

Biosimilar Monoclonal Antibody

HER2 Trastuzumab Biosimilar Monoclonal antibody specifically detects HER2 Trastuzumab Biosimilar in Human samples. It is validated for Flow cytometry|Inhibition
TRUSTED_SUSTAINABILITY

Specifications

Antigen HER2 Trastuzumab Biosimilar
Applications Flow Cytometry, Inhibition Assays
Classification Monoclonal
Clone Trastuzumab.rMAb (also known as Trastuzumab Biosimilar)
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2 μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Gene Alias ERBB2; proto-oncogene c-ErbB-2; HER-2; HER2; HER-2/neu; NEU; CD340
Immunogen A431 Cell Line Expressing EGFR
Purification Method Affinity Chromatography
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2064
Target Species Human
Content And Storage Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.