Learn More
Promega HER4 Kinase Enzyme System, 1mg
Catalog No. PRV3102 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3102 1 mg
1 options
Catalog No. PRV3102 Supplier Promega Supplier No. V3102
Add to Cart
Add to Cart
Request a Quote
Recombinant human HER4 (amino acids 682-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HER4 or ERBB4 is a transmembrane receptor tyrosine kinase that belongs to the epidermal growth factor receptor family.
  • Easily Screen and Profile HER4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C