Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Heratherm Stability Chamber 750 L

Catalog No. p-200106365
Click to view available options
Door Style:
Glass
Solid (outer), Glass (inner)
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Gain confidence in your product when testing stability and shelf life with robust Heratherm Stability chambers designed to meet a variety of quality control demands.

  • Broad relative humidity range to cover many applications, including humidity levels below ambient.
  • Environmentally friendly water-efficient humidification system.
  • Minimal condensate in standard applications.
  • Integrated water tank, fixed water connection for in-house purified water option available.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs, restrict access with standard door lock, or optional automated door lock, various alarm options to prevent operator of process issues.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Simplified setup with no technician required, install using standard power cord.
  • IQ/OQ qualifications and other services available, visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Audible and Visible
Capacity (English) 26.5 cu.ft.
Capacity (English) Shelf 66.1 lb.
Capacity (Metric) Shelf 30 kg
Certifications/Compliance UL, CE
Data Outputs USB, Ethernet
Capacity (Metric) 750 L
Chamber Material Powder-coated Cold Rolled Steel
Material Powder-coated Cold Rolled Steel
No. of Shelves 3
Plug Type Nema 5-20P
Relative Humidity 10 to 90%
Temperature Range (Metric) 0°C to 70°C
Type Humidity Chamber
Display 7 in. Touchscreen User Interface
Door Style Solid (outer), Glass (inner)
Humidity Range 10 to 90% RH
Humidity Resolution 0.1% RHNo
No. of Casters 4
Ports 2 Access Ports
Programmability Temperature and Humidity Ramping
Shelf Area (English) 4.9 ft.²
Shelf Area (Metric) 0.46 m²
Shelf Material Stainless Steel (304/1.4301)
Temperature Control Microprocessor - Programmable
Temperature Resolution 0.1°C
Temperature Stability ±0.1°C at 20°C to 37°C
Temperature Uniformity ±0.1°C at 20°C to 37°C
Warranty 24 Months
Water Consumption 0.15 to 1 L/Day
Water Resistivity 0.05 to 1 MΩ
Width (English) 42.52 in.
Width (English) Interior 33.07 in.
Width (English) Shelf 22.6 in.
Width (Metric) 1080 mm
Width (Metric) Interior 840 mm
Width (Metric) Shelf 795 mm
Height (English) 72.24 in.
Height (English) Interior 57.09 in
Height (Metric) 1835 mm
Height (Metric) Interior 1450 mm
Depth (English) 34.84 in.
Depth (English) Interior 24.8 in.
Depth (Metric) 885 mm
Depth (Metric) Interior 630 mm
Shipping Weight (English) 840 lb.
Shipping Weight (Metric) 381 kg
Electrical Requirements 100/240 V, 50/60 Hz, 1400 W
Voltage 100/115/220/230 V
Wattage 1400 W
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.