Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HERC1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105968R
Description
HERC1 Polyclonal antibody specifically detects HERC1 in Human samples. It is validated for Western BlotSpecifications
HERC1 | |
Polyclonal | |
Western Blot | |
EC 6.3.2, guanine nucleotide exchange factor p532, hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1(CHC1)-like domain (RLD) 1, hect domain and RCC1-like domain 1, HECT domain and RCC1-like domain-containing protein 1, p532probable E3 ubiquitin-protein ligase HERC1, p619EC 6.3.2.- | |
The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1 (CHC1)-like domain (RLD) 1 using the numbering given in entry NP_003913.3 (GeneID 892 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
8925 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction