Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HERC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983788
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3788 Supplier Bioss Supplier No. BS15459R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gen encodes a member of the HERC protein family. This protein stimulates guanine nucleotide exchange on ARF1 and Rab proteins. This protein may be involved in membrane transport processes.

Specifications

Antigen HERC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HERC1
Gene Accession No. Q15751
Gene Alias guanine nucleotide exchange factor p532; hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1 (CHC1)-like domain (RLD) 1; HECT and RLD domain containing E3 ubiquitin protein ligase family member 1; HECT domain and RCC1-like domain-containing protein 1; HECT-type E3 ubiquitin transferase HERC1; HERC1; p532; p619; Probable E3 ubiquitin-protein ligase HERC1
Gene Symbols HERC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HERC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8925
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.