Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HERC2 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298017G
Description
HERC2 Polyclonal specifically detects HERC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HERC2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
D15F37S1, DKFZp547P028, E3 ubiquitin-protein ligase HERC2, EC 6.3.2, EC 6.3.2.-, HECT domain and RCC1-like domain-containing protein 2, hect domain and RLD 2, jdf2, KIAA0393, p528, probable E3 ubiquitin-protein ligase HERC2, SHEP1 | |
Produced in rabbits immunized with E. coli-derived Human HERC2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
8924 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction