Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HERC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140038 Supplier Invitrogen™ Supplier No. PA5140038
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: AQVPMEYNHL QEIPIIALRN RLLLLHHLSE LFCPCIPMFD LEGSLDETGL GPSVGFDTLR GILISQGKEA AFRKVVQATM VRDRQHGPVV ELNRIQVKRS RSK Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

Highly abundant component of the nuclear matrix where it may serve a non-mitotic structural role, occupies the majority if the nuclear volume. Required for maintenance and establishment of the mitotic spindle poles, functionning as a tether linking bulk microtubules of the spindle to centrosomes. May be involved in coordination of the alignment of the mitotic spindle to the cellular polarity axis, which is a prerequisite for asymmetric cell divisions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERC2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HERC2
Gene Accession No. O95714
Gene Alias D15F32S1h; D15F37S1; D7H15F32S1; D7H15F37S1; E3 ubiquitin-protein ligase HERC2; hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1 (CHC1)-like domain (RLD) 2; HECT and RLD domain containing E3 ubiquitin protein ligase 2; hect domain and RCC1-like domain-containing protein 2; hect domain and RLD 2; HECT-type E3 ubiquitin transferase HERC2; HERC2; Jdf2; Kiaa0393; mKIAA0393; MRT38; p528; probable E3 ubiquitin-protein ligase HERC2; rjs; SHEP1
Gene Symbols HERC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HERC2. Recombinant protein control fragment (Product #RP-109391).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8924
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.