Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HERC2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15460R

Description
Highly abundant component of the nuclear matrix where it may serve a non-mitotic structural role, occupies the majority if the nuclear volume. Required for maintenance and establishment of the mitotic spindle poles, functionning as a tether linking bulk microtubules of the spindle to centrosomes. May be involved in coordination of the alignment of the mitotic spindle to the cellular polarity axis, which is a prerequisite for asymmetric cell divisions.Specifications
HERC2 | |
Polyclonal | |
Unconjugated | |
HERC2 | |
D15F32S1h; D15F37S1; D7H15F32S1; D7H15F37S1; E3 ubiquitin-protein ligase HERC2; hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1 (CHC1)-like domain (RLD) 2; HECT and RLD domain containing E3 ubiquitin protein ligase 2; HECT domain and RCC1-like domain-containing protein 2; hect domain and RLD 2; HECT-type E3 ubiquitin transferase HERC2; Herc2; Jdf2; Kiaa0393; mKIAA0393; MRT38; p528; probable E3 ubiquitin-protein ligase HERC2; Rjs; SHEP1 | |
Rabbit | |
Protein A | |
RUO | |
8924 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O95714 | |
HERC2 | |
KLH conjugated synthetic peptide derived from human HERC2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction