Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HERC3 Polyclonal Antibody
HERC3 Polyclonal Antibody
Supplier: Thermo Scientific PA576419
Description
HERC3 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
HERC3 | |
Polyclonal | |
Unconjugated | |
HERC3 | |
5730409F18Rik; AI646603; HECT and RLD domain containing E3 ubiquitin protein ligase 3; HECT domain and RCC1-like domain-containing protein 3; hect domain and RLD 3; HECT-type E3 ubiquitin transferase HERC3; HERC3; KIAA0032; mKIAA0032; Probable E3 ubiquitin-protein ligase HERC3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
73998, 8916 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15034 | |
HERC3 | |
Recombinant fusion protein of human HERC3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction