Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HERC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597119 Supplier Invitrogen™ Supplier No. PA597119
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLCWGYWSLG QPGISTNLQG IVAEPQVCGF ISDRSVKEVA CGGNHSVFLL EDGEVYTCGL NTKGQLGHER EGNKPEQIGA LADQHIIHVA CGESHSLALS DRGQLFSWGA GSDGQLGLMT TEDSVAVPRL IQKLNQQTIL QVSCGNWHCL ALAADGQFFT WGKNSHGQLG LGKEFPSQAS PQRVRSLEGI PLAQVAAGGA; Positive Samples: SH-SY5Y, U-87MG, A-431, Mouse lung, Mouse spinal cord; Cellular Location: Cytoplasm, Cytoplasmic vesicle.

This gene encodes a member the HERC ubiquitin ligase family. The encoded protein is located in the cytosol and binds ubiquitin via a HECT domain. Mutations in this gene have been associated with colorectal and gastric carcinomas. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERC3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HERC3
Gene Accession No. Q15034
Gene Alias 5730409F18Rik; AI646603; HECT and RLD domain containing E3 ubiquitin protein ligase 3; HECT domain and RCC1-like domain-containing protein 3; hect domain and RLD 3; HECT-type E3 ubiquitin transferase HERC3; HERC3; KIAA0032; mKIAA0032; Probable E3 ubiquitin-protein ligase HERC3
Gene Symbols HERC3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human HERC3 (NP_0554211).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 73998, 8916
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.