Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HERC4 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP100104AF405
Description
HERC4 Polyclonal antibody specifically detects HERC4 in Human, Mouse samples. It is validated for Western BlotSpecifications
HERC4 | |
Polyclonal | |
Western Blot | |
DKFZP564G092, EC 6.3.2, EC 6.3.2.-, HECT domain and RCC1-like domain-containing protein 4, hect domain and RLD 4, KIAA1593probable E3 ubiquitin-protein ligase HERC4 | |
The immunogen recognized by this antibody maps to a region between residue 1007 and 1057 of human hect domain and RLD 4 using the numbering given in entry NP_071362.1 (GeneID 26091). | |
0.1 mL | |
Cell Biology | |
26091 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction