Learn More
Invitrogen™ HERC4 Monoclonal Antibody (2G7)

Description
Peptide Sequence: VKGNWYPYNG QCLPDIDSEE YFCVKRIFSG GDQSFSHYSS PQNCGPPDDF RCPNPTKQIW TVNEALIQKW LSYPSGRFPV EIANEIDGTF SSSGCLNGSF.
Specifications
Specifications
| Antigen | HERC4 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2G7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Herc4 |
| Gene Accession No. | Q5GLZ8 |
| Gene Alias | 1700056O17Rik; 4921531D01Rik; 9530080M15Rik; HECT and RLD domain containing E3 ubiquitin protein ligase 4; HECT domain and RCC1-like domain-containing protein 4; hect domain and RLD 4; HECT-type E3 ubiquitin transferase HERC4; HERC4; KIAA1593; mKIAA1593; Probable E3 ubiquitin-protein ligase HERC4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.