Learn More
Invitrogen™ HERC4 Polyclonal Antibody

Description
Immunogen sequence: GNASFGQLGLG GIDEEIVLEP RKSDFFINKR VRDVGCGLRH TVFVLDDGTV YTCGCNDLGQ LGHEKSRKKP EQV Highest antigen sequence identity to the following orthologs - mouse 95%, rat 96%.
Specifications
Specifications
| Antigen | HERC4 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Herc4 |
| Gene Accession No. | Q5GLZ8 |
| Gene Alias | 1700056O17Rik; 4921531D01Rik; 9530080M15Rik; HECT and RLD domain containing E3 ubiquitin protein ligase 4; HECT domain and RCC1-like domain-containing protein 4; hect domain and RLD 4; HECT-type E3 ubiquitin transferase HERC4; HERC4; KIAA1593; mKIAA1593; Probable E3 ubiquitin-protein ligase HERC4 |
| Gene Symbols | Herc4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.