Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HERC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109808 Supplier Invitrogen™ Supplier No. PA5109808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WTVNEALIQK WLSYPSGRFP VEIANEIDGT FSSSGCLNGS FLAVSNDDHY RTGTRFSGVD MNAARLLFHK LIQPDHPQIS QQVAASLEKN LIPKLTSSLP DVEALRFYLT LPECPLMSDS NNFTTIAIPF GTALVNLEKA PLKVLENWWS VLEPPLFLKI VELFKEVVVH LLKLYKIGIP PSERRIFNSF LHTALKVLEI LHRVNEKMGQ IIQYDKFYIH EVQELIDIRN DYINWVQQQA YGMLADIPVT ICTYPFVFDA QAKTTLLQTD AVLQMQMAID QAHRQNVSSL FLPVIESVNP C.

Probable E3 ubiquitin-protein ligase involved in either protein trafficking or in the distribution of cellular structures. Required for spermatozoon maturation and fertility, and for the removal of the cytoplasmic droplet of the spermatozoon. E3 ubiquitin-protein ligases accept ubiquitin from an E2 ubiquitin- conjugating enzyme in the form of a thioester and then directly transfer it to targeted substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERC4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Herc4
Gene Accession No. Q5GLZ8, Q5PQN1, Q6PAV2
Gene Alias 1700056O17Rik; 4921531D01Rik; 9530080M15Rik; HECT and RLD domain containing E3 ubiquitin protein ligase 4; HECT domain and RCC1-like domain-containing protein 4; hect domain and RLD 4; HECT-type E3 ubiquitin transferase HERC4; HERC4; KIAA1593; mKIAA1593; Probable E3 ubiquitin-protein ligase HERC4
Gene Symbols Herc4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-700 of human HERC4 (NP_056416.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26091, 309758, 67345
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.