Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HERC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $524.00
Specifications
Antigen | HERC4 |
---|---|
Applications | Western Blot, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HERC4 Polyclonal specifically detects HERC4 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
HERC4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
26091 | |
IgG | |
Affinity purified |
Western Blot, KnockDown | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
DKFZP564G092, EC 6.3.2, EC 6.3.2.-, HECT domain and RCC1-like domain-containing protein 4, hect domain and RLD 4, KIAA1593probable E3 ubiquitin-protein ligase HERC4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 400-700 of human HERC4 (NP_056416.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title