Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HERC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542317 Supplier Invitrogen™ Supplier No. PA542317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CLVAELVGYR VTQIACGRWH TLAYVSDLGK VFSFGSGKDG QLGNGGTRDQ Sequence homology: Cow: 86%; Dog: 79%; Human: 100%.

HERC5 (E3 ISG15 - protein ligase HERC5) is a major E3 ligase for ISG15 conjugation. It acts a positive regulator of innate antiviral response in cells induced by interferon. HERC5 functions as a part of the ISGylation machinery that recognizes target protein in a broad and relatively non-specific manner. It catalyzes ISGylation of IRF3 - which results in sustained activation, it attenuates IRF3-PIN1 interaction - which antagonizes IFR3 ubiquitination and degradation, and boosts the antiviral repsonse. HERC5 is physically assocated with polyribosomes and broadly modifies newly synthesized proteins in a cotranslational manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERC5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HERC5
Gene Accession No. Q9UII4
Gene Alias CEB1; CEBP1; Cyclin-E-binding protein 1; E3 ISG15--protein ligase HERC5; HECT and RLD domain containing E3 ubiquitin protein ligase 5; HECT domain and RCC1-like domain-containing protein 5; hect domain and RLD 5; HERC5; probable E3 ubiquitin-protein ligase HERC5
Gene Symbols HERC5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HERC5 (aa 287-336).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51191
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.