Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HERC6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542322
Description
Peptide sequence: LSKDSQVFSW GKNSHGQLGL GKEFPSQASP QRVRSLEGIP LAQVAAGGAH Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%.
HERC6 belongs to the HERC family of ubiquitin ligases, all of which contain a HECT domain and at least 1 RCC1 (MIM 179710)-like domain (RLD). The 350-amino acid HECT domain is predicted to catalyze the formation of a thioester with ubiquitin before transferring it to a substrate, and the RLD is predicted to act as a guanine nucleotide exchange factor for small G proteins (Hochrainer et al., 2005 [PubMed 15676274]).
Specifications
HERC6 | |
Polyclonal | |
Unconjugated | |
HERC6 | |
1700121D12Rik; 2510038N07Rik; 4930427L17Rik; AI451296; CEB1; E3 ISG15--protein ligase Herc6; FLJ20637; HECT and RLD domain containing E3 ubiquitin protein ligase family member 6; HECT domain and RCC1-like domain-containing protein 6; hect domain and RLD 5; hect domain and RLD 6; HECT-type E3 ubiquitin transferase HERC6; Herc5; HERC6; potential ubiquitin ligase; probable E3 ubiquitin-protein ligase HERC6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55008 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IVU3 | |
HERC6 | |
Synthetic peptide directed towards the N-terminal of human HERC6 (aa 143-192). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction