Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HERC6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983793
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3793 Supplier Bioss Supplier No. BS15463R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HERC6 belongs to the HERC family of ubiquitin ligases, all of which contain a HECT domain and at least 1 RCC1 (MIM 179710)-like domain (RLD). The 350-amino acid HECT domain is predicted to catalyze the formation of a thioester with ubiquitin before transferring it to a substrate, and the RLD is predicted to act as a guanine nucleotide exchange factor for small G proteins (Hochrainer et al., 2005 [PubMed 15676274]).

Specifications

Antigen HERC6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HERC6
Gene Accession No. Q8IVU3
Gene Alias 1700121D12Rik; 2510038N07Rik; 4930427L17Rik; AI451296; BOS_6531; CEB1; E3 ISG15--protein ligase Herc6; FLJ20637; HECT and RLD domain containing E3 ubiquitin protein ligase family member 6; HECT domain and RCC1-like domain-containing protein 6; hect domain and RLD 5; hect domain and RLD 6; HECT-type E3 ubiquitin transferase HERC6; Herc5; Herc6; potential ubiquitin ligase; Probable E3 ubiquitin-protein ligase HERC6
Gene Symbols HERC6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HERC6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55008
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.