Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex I/II Monoclonal Antibody (HP12.6), QED Bioscience™
SDP

Catalog No. 502277596
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7596 1 mg
50-227-7597 100 μg
50-227-7598 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 50-227-7596 Supplier Leinco Technologies Inc Supplier No. 152031000UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Herpes Simplex I/II Monoclonal antibody specifically detects Herpes Simplex I/II in Virus samples. It is validated for ELISA, Immunocytochemistry

Specifications

Antigen Herpes Simplex I/II
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone HP12.6
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Herpes Simplex 1; Herpes Simplex 2; Herpes Simplex I; Herpes Simplex II; HHV; HHV-2; HSV; HSV 1/2; HSV I/II; HSV Type 1; HSV Type 2; HSV Type II; HSV1; HSV2; HSV-2
Host Species Mouse
Immunogen HSV1 and HSV2 infected cells
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.