Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 1 Antibody (HSV1/4055R), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB179751 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179751 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179751 Supplier Novus Biologicals Supplier No. NBP314103FR
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Herpes Simplex Virus 1 Monoclonal antibody specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Herpes Simplex Virus 1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HSV1/4055R
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1
Host Species Rabbit
Immunogen Detergent-solubilized herpes simplex virus (HSV) type 1 infected cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.