Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Herpes Simplex Virus 1 Antibody (HSVI/2045), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308639V
Description
Herpes Simplex Virus 1 Monoclonal antibody specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for Western BlotSpecifications
Herpes Simplex Virus 1 | |
Monoclonal | |
DyLight 405 | |
Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1 | |
Baculovirus-expressed Herpes Simplex Virus 1 DNA polymerase (POL) and POL/UL42 complex | |
100 μg | |
Virology Bacteria and Parasites | |
Virus | |
Purified |
Western Blot | |
HSVI/2045 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction