Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Herpes Simplex Virus 1 Antibody (HSVI/2045), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308639JF549
Description
Herpes Simplex Virus 1 (HSV1) Monoclonal antibody specifically detects Herpes Simplex Virus 1 (HSV1) in Virus samples. It is validated for Western BlotSpecifications
Herpes Simplex Virus 1 (HSV1) | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Virus | |
Purified |
Western Blot | |
HSVI/2045 | |
Western Blot | |
Herpes simplex virus 1, Herpes Simplex Virus 1 and 2, Herpes simplex virus 2, Herpes simplex virus type 1, Herpes simplex virus type 2, HSV 1, HSV 2 | |
Baculovirus-expressed HSV DNA polymerase (POL) and POL/UL42 complex | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction