Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Herpes Simplex Virus 1 Antibody (T111), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500476AF532
Description
Herpes Simplex Virus 1 Monoclonal antibody specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence.Specifications
Herpes Simplex Virus 1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
Recognizes gB antigen of both Herpes Simplex Virus 1 &2 | |
Store at 4°C in the dark. |
ELISA, Immunocytochemistry, Immunofluorescence | |
T111 | |
ELISA, Immunocytochemistry/Immunofluorescence, Immunocytochemistry | |
Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1 | |
Exctract of Herpes Simplex Virus 1 infected VERO (green monkey kidney) cells. | |
Primary | |
Virus | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction