Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Herpes Simplex Virus 1 Antibody (T96) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500477
Description
Herpes Simplex Virus 1 Monoclonal specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Herpes Simplex Virus 1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, ELISA 1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Virus | |
IgG2b |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
T96 | |
Unconjugated | |
Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1 | |
Exctract of HSV-1 infected VERO (green monkey kidney) cells. | |
0.1 mg | |
Virology Bacteria and Parasites | |
The antibody T96 recognizes gC antigen of HSV1 (Herpes Simplex Virus type 1), a dsDNA virus, member of Herpesviridae family. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction