Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Herpes Simplex Virus 2 Antibody (T303), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500478AF488
Description
Herpes Simplex Virus 2 Monoclonal antibody specifically detects Herpes Simplex Virus 2 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.Specifications
Herpes Simplex Virus 2 | |
Monoclonal | |
Alexa Fluor 488 | |
Herpes Simplex Virus Type 2 | |
Exctract of Herpes Simplex Virus 2 infected VERO (green monkey kidney) cells. | |
0.1 mL | |
Primary | |
Virus | |
IgG2a |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
T303 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Affinity Purified | |
RUO | |
The antibody T303 recognizes gG antigen of HSV2 (Herpes Simplex Virus type 2), a dsDNA virus, member of Herpesviridae family. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction