Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus Glycoprotein D Mouse anti-Virus, Unconjugated, Clone: , MilliporeSigma™

Catalog No. MABF197525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1975-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1975-25 Supplier MilliporeSigma Supplier No. MABF197525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Herpes Simplex Virus Glycoprotein D, clone LP14, Cat. No. MABF1975, is a mouse monoclonal antibody that detects Glycoprotein D of Herpes Simplex Virus and has been tested for use in Inhibition, Immunocytochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Herpes Simplex Virus Glycoprotein D
Applications Inhibition Assays, Immunocytochemistry, Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in PBS without azide.
Gene Alias HSV glycoprotein D
Gene Symbols N/A
Host Species Mouse
Immunogen Whole Herpex Simplex Virus 2 (HSV-2).
Molecular Weight of Antigen ∽50 and 55 kDa observed; 43.35 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone LP14 detects glycoprotein D of Herpes Simplex virus.
Target Species Virus
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.