Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Herpes Simplex Virus Type 1/2 gD Monoclonal Antibody (A18S), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518211
Description
This antibody reacts with HSV type 1 and 2, glycoprotein D.
Members of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer's disease.
Specifications
Herpes Simplex Virus Type 1/2 gD | |
Monoclonal | |
100 μg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
A18S | |
FITC | |
Herpes Simplex 1; Herpes Simplex 2; Herpes Simplex I; Herpes Simplex II; HHV; HHV-2; HSV; HSV 1/2; HSV I/II; HSV Type 1; HSV Type 2; HSV Type II; HSV1; HSV2; HSV-2 | |
Herpes simplex virus, 1 & 2 native protein. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction