Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes simplex virus type 2 gC-2 Mouse anti-Herpes simplex virus, Clone: P1111, Abnova™

Catalog No. 89033549 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-549 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-549 Supplier Abnova Corporation Supplier No. MAB13586
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against herpes simplex virus type 2 (HSV-2) gC-2.

  • High specificity
  • High reactivity

Specifications

Antigen Herpes simplex virus type 2 gC-2
Applications Immunofluorescence
Classification Monoclonal
Clone P1111
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against herpes simplex virus type 2 (HSV-2) gC-2.
Dilution Immunofluorescence The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4.
Host Species Mouse
Purification Method Protein G purification
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with gC-2 of herpes simplex virus type 2.
Target Species Herpes Simplex virus (HSV)
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.