Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Herpes simplex virus type 2 gE Mouse anti-Herpes simplex virus, Clone: H2A055, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB13587
Description
Mouse monoclonal antibody raised against herpes simplex virus type 2 (HSV-2) gE.
- High specificity
- High reactivity
Specifications
Herpes simplex virus type 2 gE | |
Monoclonal | |
Unconjugated | |
ELISA Immunofluorescence The optimal working dilution should be determined by the end user. | |
Mouse | |
RUO | |
This antibody reacts with gE of herpes simplex virus type 2. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |
ELISA, Immunofluorescence | |
H2A055 | |
Mouse monoclonal antibody raised against herpes simplex virus type 2 (HSV-2) gE. | |
In PBS, pH 7.4. | |
100 μg | |
Primary | |
Herpes Simplex virus (HSV) | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction