Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus Type 6A (strain Uganda-1102) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5103298
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103298 Supplier Thermo Scientific Supplier No. PA5103298

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GCVK_HHV6U.

Human herpesvirus 6 is the common collective name for Human beta herpesvirus 6A and Human beta herpesvirus 6B. These closely related viruses are two of the nine herpesviruses known to have humans as their primary host.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Herpes Simplex Virus Type 6A (strain Uganda-1102)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Host Species Rabbit
Immunogen A synthesized peptide derived from human virus GCVK_HHV6U, corresponding to a region within C-terminal amino acids.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.