Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Herpes Simplex Virus UL42 Antibody (13C9), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002761G
Description
HSV UL42 Monoclonal specifically detects HSV UL42 in Virus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
UL42 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
13C9 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
HHV1gp061 | |
Baculovirus-expressed HSV DNA polymerase (POL) and POL/UL42 complex. | |
0.1 mL | |
Virology Bacteria and Parasites | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction