Learn More
HERPUD2 Rabbit anti-Human, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 206401AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-3008
Only null left
Add to Cart
Description
Description
Could be involved in the unfolded protein response (UPR) pathway.
Specifications
Specifications
HERPUD2
Polyclonal
Unconjugated
HERPUD2
HERPUD family member 2, HERPUD2
Rabbit
Antigen Affinity Chromatography
RUO
64224
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BSE4
HERPUD2
HERPUD2 Fusion Protein Ag14702
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.