Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HERV-FRD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112876 Supplier Invitrogen™ Supplier No. PA5112876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HERV-FRD Polyclonal Antibody for Western Blot, ELISA

Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. This endogenous envelope protein has retained its original fusogenic properties. HERV-FRD can make pseudotypes with MLV, HIV-1 or SIV-1 virions and confer infectivity.Human endogenous retroviruses (HERVs) make up approximately 8% of the human genome. Although most HERVs are nonfunctional, the HERV-W (ERVWE1; MIM 604659) and HERV-FRD envelope (env) proteins can induce cell-cell fusion when expressed in cells possessing appropriate receptors (Blaise et al., 2003 [PubMed 14557543]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERV-FRD
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ERVFRD-1
Gene Accession No. P60508
Gene Alias Endogenous retrovirus group FRD member 1; endogenous retrovirus group FRD, member 1; envelope glycoprotein syncytin-A; envelope polyprotein; envFRD; ERVFRD-1; ERVFRDE1; FLJ41944; FLJ90611; GLLL6191; Gm52; HERV-FRD; HERV-FRD provirus ancestral Env polyprotein; HERV-FRD_6p24.1 provirus ancestral Env polyprotein; HERV-W/FRD; MGC87585; SU; Surface protein; Syna; Synb; syncytin 2; syncytin a; Syncytin2; syncytin-2; TM; Transmembrane protein; UNQ6191; UNQ6191/PRO20218
Gene Symbols ERVFRD-1
Host Species Rabbit
Immunogen Recombinant Human Syncytin-2 protein (16-250AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 405754
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.