Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HERV Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595785
Description
Positive Samples: U-251MG, 293T, mouse brain; Cellular Location: Cell membrane, Peripheral membrane protein, Single-pass type I membrane protein, Virion Immunogen sequence: PEQWNNFSTE INTTS.
HERVs are a class of multiple sclerosis associated retroviruses. Reacts with a replication competent Human Endogenous Retrovirus (HERVW) isolated from placenta. The synthetic peptide sequence used as immunogen is taken from the virus envelope polyprotein sequence. Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. The HERVW endogenous envelope protein has retained its original fusogenic properties and participates in trophoblast fusion during placenta morphogenesis.
Specifications
HERV | |
Polyclonal | |
Unconjugated | |
ERVW-1 | |
endogenous retroviral family W, env(C7), member 1; endogenous retrovirus group W member 1; endogenous retrovirus group W, member 1; ENV; envelope glycoprotein; envelope polyprotein gPr73; envelope protein; Enverin; ENVW; Env-W; ERVW-1; ERVWE1; gp24; gp50; HERV7Q; HERV-7q; HERV-7q envelope protein; HERV-tryptophan envelope protein; HERVW; HERV-W Env glycoprotein; HERV-W envelope protein; HERV-W_7q21.2 provirus ancestral Env polyprotein; HERVWENV; HERV-W-ENV; human endogenous retrovirus W envC7-1 envelope protein; SU; Surface protein; Syncytin; syncytin A; syncytin-1; TM; Transmembrane protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
30816 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.16 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q9UQF0 | |
ERVW-1 | |
A synthetic peptide corresponding to a sequence within amino acids 201-300 of human ERVW-1 (NP_0554053). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction