Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HERV Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA512286
Description
HERV Polyclonal Antibody for Western Blot, IHC (P)
HERVs are a class of multiple sclerosis associated retroviruses. Reacts with a replication competent Human Endogenous Retrovirus (HERVW) isolated from placenta. The synthetic peptide sequence used as immunogen is taken from the virus envelope polyprotein sequence. Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. The HERVW endogenous envelope protein has retained its original fusogenic properties and participates in trophoblast fusion during placenta morphogenesis.
Specifications
HERV | |
Polyclonal | |
Unconjugated | |
ERVW-1 | |
endogenous retroviral family W, env(C7), member 1; endogenous retrovirus group W member 1; endogenous retrovirus group W, member 1; ENV; envelope glycoprotein; envelope polyprotein gPr73; envelope protein; Enverin; ENVW; Env-W; ERVW-1; ERVWE1; gp24; gp50; HERV7Q; HERV-7q; HERV-7q envelope protein; HERV-tryptophan envelope protein; HERVW; HERV-W Env glycoprotein; HERV-W envelope protein; HERV-W_7q21.2 provirus ancestral Env polyprotein; HERVWENV; HERV-W-ENV; human endogenous retrovirus W envC7-1 envelope protein; SU; Surface protein; Syncytin; syncytin A; syncytin-1; TM; Transmembrane protein | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
30816 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9UQF0 | |
ERVW-1 | |
KLH conjugated synthetic peptide between 400-429 amino acids from the central region of human HERV (ERVWE1). | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction